Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%
Acros Organics

Tetraphenylboron sodium, 99.5%

Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g | CAS: 143-66-8 | C24H20BNa | 342.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 138521000
HSN 98020000
Price (INR)
40,099.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
40,099.00
Each
Chemical Identifiers
CAS143-66-8
IUPAC Namesodium tetraphenylboranuide
Molecular FormulaC24H20BNa
InChI KeyHFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO499.4 to 100.6 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only