Thenoyltrifluoroacetone, 99%
Thenoyltrifluoroacetone, 99%
Thenoyltrifluoroacetone, 99%
Acros Organics

Thenoyltrifluoroacetone, 99%

CAS: 326-91-0 | C8H5F3O2S | 222.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 138611000
also known as 13861
HSN 32049000
Price (INR)
22,914.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
22,914.00
Each
Chemical Identifiers
CAS326-91-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Light yellow to beige to brown
Melting point40°C to 44°C
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only