2-Thiophenecarboxylic acid, 99%
2-Thiophenecarboxylic acid, 99%
2-Thiophenecarboxylic acid, 99%
Acros Organics

2-Thiophenecarboxylic acid, 99%

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number 138841000
also known as 13884
HSN 29349900
Price (INR)
7,741.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
7,741.00
Each
Chemical Identifiers
CAS527-72-0
IUPAC Namethiophene-2-carboxylic acid
Molecular FormulaC5H4O2S
InChI KeyQERYCTSHXKAMIS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to cream to light brown
Melting point126°C to 130°C
GC>=98.5 %
Titration with NaOH98.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only