2-Thiophenecarboxylic acid hydrazide, 97%
2-Thiophenecarboxylic acid hydrazide, 97%
2-Thiophenecarboxylic acid hydrazide, 97%
Acros Organics

2-Thiophenecarboxylic acid hydrazide, 97%

CAS: 2361-27-5 | C5H6N2OS | 142.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 138850050
also known as 13885
HSN 29349900
Price (INR)
8,214.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,214.00
Each
Chemical Identifiers
CAS2361-27-5
IUPAC Namethiophene-2-carbohydrazide
Molecular FormulaC5H6N2OS
InChI KeySOGBOGBTIKMGFS-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point136°C to 140°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only