o-Toluic acid, 98+%
o-Toluic acid, 98+%
o-Toluic acid, 98+%
Acros Organics

o-Toluic acid, 98+%

CAS: 118-90-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number 139040050
also known as 13904
HSN 29089990
Price (INR)
2,533.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
2,533.00
Each
Chemical Identifiers
CAS118-90-1
IUPAC Name2-methylbenzoic acid
Molecular FormulaC8H8O2
InChI KeyZWLPBLYKEWSWPD-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point102°C to 106°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
Titration with NaOH>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only