Tricarballylic acid, 99%
Tricarballylic acid, 99%
Tricarballylic acid, 99%
Acros Organics

Tricarballylic acid, 99%

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol
Have Questions?
Catalog NumberQuantity
139360500
also known as 13936
50 g
Catalog number 139360500
also known as 13936
HSN 29171990
Price (INR)
25,305.00
Each
Quantity:
50 g
Request bulk or custom format
Price (INR)
25,305.00
Each
Chemical Identifiers
CAS99-14-9
IUPAC Namepropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O6
InChI KeyKQTIIICEAUMSDG-UHFFFAOYSA-N
SMILESOC(=O)CC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point154°C to 159°C
Titration with TBAH>=98.5 %
Appearance (Color)Off-white to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only