n-Tridecane, 99+%
n-Tridecane, 99+%
n-Tridecane, 99+%
Acros Organics

n-Tridecane, 99+%

CAS: 629-50-5 | C13H28 | 184.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 139510250
HSN 29011000
Price (INR)
4,166.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,166.00
Each
Chemical Identifiers
CAS629-50-5
IUPAC Nametridecane
Molecular FormulaC13H28
InChI KeyIIYFAKIEWZDVMP-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only