Triethyl 1,1,2-ethanetricarboxylate, 99%
Triethyl 1,1,2-ethanetricarboxylate, 99%
Triethyl 1,1,2-ethanetricarboxylate, 99%
Acros Organics

Triethyl 1,1,2-ethanetricarboxylate, 99%

CAS: 7459-46-3 | C11H18O6 | 246.259 g/mol
Have Questions?
Catalog NumberQuantityPackaging
139610250
also known as 13961
25 gGlass bottle
Catalog number 139610250
also known as 13961
HSN 29171990
Price (INR)
7,149.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,149.00
Each
Chemical Identifiers
CAS7459-46-3
IUPAC Name1,1,2-triethyl ethane-1,1,2-tricarboxylate
Molecular FormulaC11H18O6
InChI KeyTVWZLLYAJDSSCJ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=98.5 %
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only