Triethyl phosphonoacetate, 97%
Triethyl phosphonoacetate, 97%
Triethyl phosphonoacetate, 97%
Acros Organics

Triethyl phosphonoacetate, 97%

CAS: 867-13-0 | C8H17O5P | 224.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 139701000
HSN 98020000
Price (INR)
9,563.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,563.00
Each
Chemical Identifiers
CAS867-13-0
IUPAC Nameethyl 2-(diethoxyphosphoryl)acetate
Molecular FormulaC8H17O5P
InChI KeyGGUBFICZYGKNTD-UHFFFAOYSA-N
SMILESCCOC(=O)CP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Appearance (Form)Clear liquid
Acid value=<2 mg KOH/g
GC>=96.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only