Trifluoroacetic acid, silver salt, 98%
Trifluoroacetic acid, silver salt, 98%
Trifluoroacetic acid, silver salt, 98%
Acros Organics

Trifluoroacetic acid, silver salt, 98%

CAS: 2966-50-9 | C2AgF3O2 | 220.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 139730250
also known as 13973
HSN 28432900
Price (INR)
32,122.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
32,122.00
Each
Chemical Identifiers
CAS2966-50-9
IUPAC Namesilver(1+) trifluoroacetate
Molecular FormulaC2AgF3O2
InChI KeyKZJPVUDYAMEDRM-UHFFFAOYSA-M
SMILES[Ag+].[O-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Light beige to grey
Infrared spectrumConforms
Titration with NH4SCN>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only