Product Image
Thermo Scientific™

DRI™ THC (Cannabinoid) Drugs of Abuse Controls

Eliminate time-consuming steps in reagent preparation with the Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls—ready-to-use controls in liquid form.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
14011 x 25 mL40 ng/mL
01681 x 5 mL60 ng/mL
01701 x 5 mL40 ng/mL
02121 x 5 mL125 ng/mL
14021 x 25 mL60 ng/mL
14041 x 25 mL125 ng/mL
02141 x 5 mL75 ng/mL
Catalog number 1401
Price (INR)
-
Quantity:
1 x 25 mL
Concentration:
40 ng/mL
Validate the Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Assay and ensure accuracy of results with Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls. These liquid, ready-to-use controls streamline your testing process.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration40 ng/mL
DescriptionDRI™ THC (Cannabinoid) Drugs of Abuse Control 40ng/mL, 25mL
Quantity1 x 25 mL
TypeControl
Detectable Analytes11-nor-9-COOH-D9-THC
Sample TypeUrine
Unit SizeEach