DPBS, calcium, magnesium
Actual product may vary
DPBS, calcium, magnesium
DPBS, calcium, magnesium
Gibco™

DPBS, calcium, magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityOsmolality
14040166
also known as 14040-166
20 x 100 mL270 to 310 mOsm/kg
14040083
also known as 14040-083
100 mL270 to 310 mOsm/kg
1404018210 x 500 mL270 to 300 mOsm/kg
14040141100 mL270 to 300 mOsm/kg
14040133500 mL270 to 300 mOsm/kg
14040091
also known as 14040-091
500 mL270 to 310 mOsm/kg
14040174
also known as 14040-174
10 x 500 mL270 to 310 mOsm/kg
140401171000 mL270 to 300 mOsm/kg
140402166 x 1000 mL270 to 300 mOsm/kg
Catalog number 14040166
also known as 14040-166
Price (INR)
-
Quantity:
20 x 100 mL
Osmolality:
270 to 310 mOsm/kg
Customize this product

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 to 310 mOsm/kg
Recommended StorageStorage Conditions: 2°C to 8°C, Shipping Conditions: Ambient, Shelf Life: 36 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents
Physical FormLiquid
Product LineGibco
Quantity20 x 100 mL
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.0 to 7.2
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.