DRI™ Alcohol Negative Drugs of Abuse Calibrators
Actual product may vary
Thermo Scientific™

DRI™ Alcohol Negative Drugs of Abuse Calibrators

Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14051 x 25 mL
03111 x 5 mL
Catalog number 1405
Price (INR)
-
Quantity:
1 x 25 mL
Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy. Ready-to-use in liquid form, Thermo Scientific™ DRI™ Alcohol Negative Drugs of Abuse Calibrators have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0 ng/mL
DescriptionDRI Alcohol Negative Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 25 mL
TypeCalibrator
Detectable AnalytesAlcohol
Sample TypePlasma, Serum, Urine
Unit SizeEach
Contents & Storage
Do not Freeze; 2° to 8°C