Nunc™ Cell Factory™ Racks and Carts
Actual product may vary
Thermo Scientific™
Nunc™ Cell Factory™ Racks and Carts
Hold up to four Cell Factory 40 or 12 Cell Facotry 10 systems with Nunc Cell Factory Racks and Carts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
140503Side Loading Rack with CartCell Factory 40
120289Retrofit Clamping KitShaker
140504Side Loading Rack with CartCell Factory 10
Catalog number 140503
Price (INR)
-
Product Type:
Side Loading Rack with Cart
For Use With (Equipment):
Cell Factory 40
Store and transport equipment effectively using Thermo Scientific™ Nunc™ Cell Factory™ Racks and Carts, which can hold up to four Cell Factory 40 or 12 Cell Factory 10 systems. These racks and carts are designed to integrate perfectly with all Nunc Cell Factory equipment.
  • Constructed of AISI 316 Stainless Steel
  • Designed for use in clean room environments
  • Side-loading design for easy loading and unloading
Specifications
DescriptionSide Loading Rack with Cart for Cell Factory 40
For Use With (Equipment)Cell Factory 40
Product TypeSide Loading Rack with Cart
MaterialStainless Steel (AISI 316)
Unit SizeEach