L-Tyrosine methyl ester, 98%
L-Tyrosine methyl ester, 98%
L-Tyrosine methyl ester, 98%
Acros Organics

L-Tyrosine methyl ester, 98%

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 140650250
also known as 14065
HSN 29225090
Price (INR)
14,866.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,866.00
Each
Chemical Identifiers
CAS1080-06-4
IUPAC Namemethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC10H13NO3
InChI KeyMWZPENIJLUWBSY-VIFPVBQESA-N
SMILESCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=97.5 %
Specific optical rotation+23° to +29° (20°C, 589 nm) (c=2.4, CH3OH)
Appearance of solution(5% soln. in methanol) Clear to slightly hazy
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only