Product Image
Gibco™

DPBS (10X), calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14080048
also known as 14080-048
500 mL
14080089
also known as 14080-089
10 x 500 mL
Catalog number 14080048
also known as 14080-048
Price (INR)
-
Quantity:
500 mL
Customize this product

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

The complete formulation is available.

Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality2700 - 3200 mOsm/kg
Recommended StorageStorage conditions: 2-8° C.
Shelf life: 24 months from date of manufacture.
SterilitySterile-filtered
Concentration10 X
For Use With (Application)Mammalian Cell Culture
Physical FormLiquid
Product LineGibco
Quantity500 mL
Solution TypeDulbecco's Phosphate Buffered Saline
pH4.3 - 4.7
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.