Nunc™ Cell Factory™ System Accessories
Thermo Scientific™
Nunc™ Cell Factory™ System Accessories
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
140815Tubing Set, Female MPCCell Factory Systems (All)
140099TSPC ConnectorCell Factory Systems (All)
140050FunnelCell Factory Systems (All)
167530Filter Adapter CapCell Factory Systems (All)
170615Cover CapCell Factory Systems (All)
179553Airvent FilterCell Factory Systems (All)
140067Filter Assembly, 37 mmEasyFill Cell Factory Systems
147074Vent/Close CapCell Factory Systems (All)
173250FEP ConnectorCell Factory Systems (All)
Catalog number 140815
Price (INR)
27,028.00
Online offer
Ends: 31-Dec-2026
30,031.00
Save 3,003.00 (10%)
Case of
Product Type:
Tubing Set, Female MPC
For Use With (Equipment):
Cell Factory Systems (All)
Price (INR)
27,028.00
Online offer
Ends: 31-Dec-2026
30,031.00
Save 3,003.00 (10%)
Case of
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections made easy
  • Plug a media bag or tubing set into any cell factory
Save time and reduce costs
  • Cleaning processes and sterility validations are minimized
  • Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
Prevent buildup of back pressure during filling application
  • Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTubing Set with Female MPC connector
For Use With (Equipment)Cell Factory Systems (All)
No. per Case2
No. per Pack1
Product TypeTubing Set, Female MPC
SterilitySterile
Unit SizeCase of