Pump, 2 x 10 cc 650 μL DM (for the 140C instrument)
Product Image
Thermo Scientific™

Pump, 2 x 10 cc 650 μL DM (for the 140C instrument)

This is a replacement part for the 140C Instrument.For Research Use Only. Not for use in diagnostics procedures.Read more
Have Questions?
Catalog NumberQuantity
140C00
also known as 140C-00
1 U
Catalog number 140C00
also known as 140C-00
Price (INR)
-
Quantity:
1 U
This is a replacement part for the 140C Instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 U
Shipping ConditionRoom Temperature
Product TypePump
Unit SizeEach
Contents & Storage
Store at room temperature.