Product Image
Gibco™

HBSS, no calcium, no magnesium, no phenol red

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14175046100 mL
14175053
also known as 14175-053
500 mL
14175129
also known as 14175-129
10 x 500 mL
14175137
also known as 14175-137
20 x 100 mL
Catalog number 14175046
Price (INR)
-
Quantity:
100 mL
Customize this product

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This HBSS is modified as follows:

With: Glucose, Sodium Bicarbonate

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialHanks' Balanced Salt Solution (HBSS)
IncludesNo Calcium, No Magnesium
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 - 310 mOsm/kg
Recommended StorageStorage conditions: 15-30° C.
Shipping conditions: Ambient.
Shelf life: 36 months from date of manufacture.
SterilitySterile-filtered
Concentration1 X
Physical FormLiquid
Product LineGibco
Quantity100 mL
Solution TypeHank's Balanced Salt
pH6.7 - 7.8
Unit SizeEach

Frequently asked questions (FAQs)

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.