DPBS, no calcium, no magnesium
Gibco™

DPBS, no calcium, no magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
14190240
also known as 14190-240
10 LBag
14190086
also known as 14190-086
100 mLBottle
14190185
also known as 14190-185
20 x 100 mLBottle
14190144500 mLBottle
1419025010 x 500 mLBottle
14190169
also known as 14190-169
10 x 500 mLBottle
141901361000 mLBottle
141902356 x 1000 mLBottle
141903425 LBag
1419035910 LBag
1419036720 LBag
Catalog number 14190240
also known as 14190-240
Price (INR)
-
Quantity:
10 L
Packaging:
Bag
Customize this product

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Calcium, No Magnesium
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 300 mOsm/kg
PackagingBag
Recommended StorageStorage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Mammalian Cell Culture
Physical FormLiquid
Product LineGibco, SynerGy
Quantity10 L
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.0 to 7.3
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.