Multiskan FC is discontinued. Upgrade to Multiskan Ease with larger touchscreen operation, long-life LEDs, and durable hard-coated filters.
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Thermo Scientific™
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeWavelength
1428205Optical Filter820 nm
1424925Optical Filter492 nm
1425305Optical Filter530 nm
1425605Optical Filter560 nm
1427405Optical Filter740 nm
Catalog number 1428205
Price (INR)
72,084.00
線上優惠
Ends: 31-Dec-2026
120,140.00
Save 48,056.00 (40%)
Each
Product Type:
Optical Filter
Wavelength:
820 nm
Price (INR)
72,084.00
線上優惠
Ends: 31-Dec-2026
120,140.00
Save 48,056.00 (40%)
Each
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Product LineMultiskan
Wavelength820 nm
For Use With (Equipment)Multiskan FC
Product TypeOptical Filter
Unit SizeEach