Cytochrome C, 90%, from horse heart muscle
Acros Organics

Cytochrome C, 90%, from horse heart muscle

Cytochrome C, CAS # 9007-43-6, is a component of the electron transport chain in mitochondria, making it an important electron transporter that forms an electron bridge between respirable substrates and oxygen.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100mg
1g
Catalog number 147531000
also known as 14753
HSN 98020000
Price (INR)
16,168.00
Each
Quantity:
100mg
Request bulk or custom format
Price (INR)
16,168.00
Each
Chemical Identifiers
CAS9007-43-6
IUPAC Name(1Z,6Z,17Z)-15,19-bis(2-carboxyethyl)-4,9-bis[1-(cysteinylsulfanyl)ethyl]-5,10,14,20-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,14,16(24),17,19-decaene-22,22-bis(ylium)
Molecular FormulaC40H48FeN6O6S4
InChI KeyUMUOLVQYJUIZKD-RHANTFMNSA-L
SMILESCC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C
View more
SpecificationsSpecification SheetSpecification Sheet
IdentificationPasses test
Appearance (Color)Red-brown to dark brown
Ratio of Absorbances>=1.0 (A 550 red/A 278 ox)
UV spectrophotometry>=90 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Cytochrome C is a key player of the electron transport chain in mitochondria
• It plays an important role in apoptotic pathways
• It also plays a role in cellular oxidation in animals and plants

which make it an effective biological electron carrier. It plays a vital role in cellular oxidation in animals and plants.

Applications

• Cytochrome c shows peroxidase activity by oxidation of various electron donors such as 2,2-azino-bis (3-ethylbenzthiazoline-6-sulphonic acid) (ABTS), 2-keto-4-thiomethyl butyric acid and 4-aminoantipyrine

• It is assumed to be the functional complex utilized in low-level laser therapy (LLLT), which increases the metabolic activity and frees up more energy for the cells to repair the tissue

• It is used as an important mediator in apoptotic pathways

RUO – Research Use Only

General References:

  1. Li, J.; Sun, R.; Hao, C.; He, G.; Zhang, L.; Wang, J. The behavior of the adsorption of cytochrome C on lipid monolayers: A study by the Langmuir-Blodgett technique and theoretical analysis. Biophys. Chem. 2015, 205, 33-40.
  2. Choi, J.; Cho, D. W.; Tojo, S.; Fujitsuka, M.; Majim, T. Configurational changes of heme followed by cytochrome c folding reaction. Mol. BioSyst. 2015, 11 (1), 218-222.