Methyl 1-methylcyclopropyl ketone, 95%
Methyl 1-methylcyclopropyl ketone, 95%
Methyl 1-methylcyclopropyl ketone, 95%
Acros Organics

Methyl 1-methylcyclopropyl ketone, 95%

CAS: 1567-75-5 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 147630050
also known as 14763
HSN 29142990
Price (INR)
12,404.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,404.00
Each
Chemical Identifiers
CAS1567-75-5
IUPAC Name1-(1-methylcyclopropyl)ethan-1-one
Molecular FormulaC6H10O
InChI KeyOQBCJXUAQQMTRW-UHFFFAOYSA-N
SMILESCC(=O)C1(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4320 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only