1-Adamantanecarboxylic acid, 99%
1-Adamantanecarboxylic acid, 99%
1-Adamantanecarboxylic acid, 99%
Acros Organics

1-Adamantanecarboxylic acid, 99%

CAS: 828-51-3 | C11H16O2 | 180.25 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1478800505 gGlass Bottle
Catalog number 147880050
HSN 29162000
Price (INR)
3,148.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,148.00
Each
Chemical Identifiers
CAS828-51-3
IUPAC Nameadamantane-1-carboxylic acid
Molecular FormulaC11H16O2
InChI KeyJIMXXGFJRDUSRO-UHFFFAOYSA-N
SMILESOC(=O)C12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point172°C to 176°C
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only