Formic acid, 98+%, pure
Formic acid, 98+%, pure
Formic acid, 98+%, pure
Acros Organics

Formic acid, 98+%, pure

CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic bottle
Catalog number 147932500
HSN 29151100
Price (INR)
4,805.00
Each
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,805.00
Each
Chemical Identifiers
CAS64-18-6
IUPAC Nameformic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-UHFFFAOYSA-N
SMILESOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Color scale=<20 APHA
Refractive index1.3700 to 1.3720 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only