4-Nitro-o-phenylenediamine, 98%
4-Nitro-o-phenylenediamine, 98%
4-Nitro-o-phenylenediamine, 98%
Acros Organics

4-Nitro-o-phenylenediamine, 98%

CAS: 99-56-9 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
148845000
also known as 14884
500 gGlass bottle
Catalog number 148845000
also known as 14884
HSN 29215130
Price (INR)
72,387.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
72,387.00
Each
Chemical Identifiers
CAS99-56-9
IUPAC Name4-nitrobenzene-1,2-diamine
Molecular FormulaC6H7N3O2
InChI KeyRAUWPNXIALNKQM-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)Red-brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only