Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Acros Organics

Methyl thioglycolate, 98%

CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
148881000100 gGlass bottle
Catalog number 148881000
HSN 29309099
Price (INR)
4,308.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,308.00
Each
Add to cart
Chemical Identifiers
CAS2365-48-2
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4640 to 1.4670 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only