2-Amino-6-chloropurine, 99+%, Thermo Scientific Chemicals
2-Amino-6-chloropurine, 99+%, Thermo Scientific Chemicals
2-Amino-6-chloropurine, 99+%, Thermo Scientific Chemicals
Acros Organics

2-Amino-6-chloropurine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 149290050
also known as 14929
HSN 29335990
Price (INR)
19,695.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
19,695.00
Each
Chemical Identifiers
CAS10310-21-1
IUPAC Name6-chloro-7H-purin-2-amine
Molecular FormulaC5H4ClN5
InChI KeyRYYIULNRIVUMTQ-UHFFFAOYSA-N
SMILESNC1=NC(Cl)=C2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
HPLC>=99.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only