1,6-Heptadien-4-ol, 97%
1,6-Heptadien-4-ol, 97%
1,6-Heptadien-4-ol, 97%
Acros Organics

1,6-Heptadien-4-ol, 97%

CAS: 2883-45-6 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 150090250
also known as 15009
HSN 29052900
Price (INR)
75,749.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
75,749.00
Each
Chemical Identifiers
CAS2883-45-6
IUPAC Namehepta-1,6-dien-4-ol
Molecular FormulaC7H12O
InChI KeyUTGFOWQYZKTZTN-UHFFFAOYSA-N
SMILESOC(CC=C)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.4490 to 1.4530 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only