Nunc™ 14mL Round-Bottom Tube
Nunc™ 14mL Round-Bottom Tube
Actual product may vary
Nunc™ 14mL Round-Bottom Tube
Nunc™ 14mL Round-Bottom Tube
Thermo Scientific™

Nunc™ 14mL Round-Bottom Tube

Perform critical cellular applications with these 14mL round-bottom tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. per CaseNo. per Pack
1502854001
15026850010
Catalog number 150285
Price (INR)
-
No. per Case:
400
No. per Pack:
1
Perform critical cellular applications with Thermo Scientific™ Nunc™ 14mL Round-Bottom Tubes, facilitated by the dual position vent/close cap for optimized gas exchange. For research use only
  • Optimized gas exchange with dual position vent/close cap
  • Encased in breathable packaging
  • Certificates of conformity available for every lot
  • For research use only
Specifications
Certifications/ComplianceMouse Embryo Assay (1-cell stage MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Non-pyrogenicand sterile (SAL 10-6).
Closure MaterialLDPE
Diameter (Metric)17.5 mm
No. per Case400
No. per Pack1
ShapeRound
Capacity (Metric)14 mL
Closure TypeDual Position Vent/Close Cap
Length (Metric)95 mm
MaterialPolystyrene
Max. RCF6000 x g
Product TypeRound Bottom Tube
SterilitySterile
Unit SizeCase of 400