Pierce™ Maleic Anhydride Activated Plates
Pierce™ Maleic Anhydride Activated Plates
Pierce™ Maleic Anhydride Activated Plates
Thermo Scientific™
Pierce™ Maleic Anhydride Activated Plates
Thermo Scientific™ Pierce Maleic Anhydride Activated Plates allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFormatQuantity
15102ClearStrip Plate5 x 5 Plates
15100ClearStrip Plate5 Plates
15110ClearStandard5 Plates
15112ClearStandard5 x 5 Plates
Catalog number 15102
Price (INR)
103,424.00
Online offer
Ends: 31-Mar-2026
172,372.00
Save 68,948.00 (40%)
Each
Color:
Clear
Format:
Strip Plate
Quantity:
5 x 5 Plates
Request bulk or custom format
Price (INR)
103,424.00
Online offer
Ends: 31-Mar-2026
172,372.00
Save 68,948.00 (40%)
Each
Pierce Maleic Anhydride Activated Plates are useful for immobilizing peptides and other ligands that do not coat efficiently by passive adsorption. Reaction of the maleic anhydride groups with primary amines (–NH2) results in formation of amide bonds that are stable at neutral pH and above. Acidic conditions hydrolyze the bond; therefore, coating reactions are best performed at pH 8 to 9, and the reacted plates are best used at pH greater than 7 for ELISA and other methods.
These Thermo Scientific Pierce Maleic Anhydride Activated 96-Well Plates are clear, 96-well plates that allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.
Features of Maleic Anhydride Activated Plates:
• Spontaneously react with primary amines
• A direct, simple approach to covalently attach molecules to the well surface of an EIA plate
• Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Covalent
Detection MethodColorimetric
No. per Pack5 Plates
Plate BlockingNone
Product TypeDetection Plate, Immunoassay, ELISA
Quantity5 x 5 Plates
ColorClear
FormatStrip Plate
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentMaleic Anhydride
Unit SizeEach
Contents & Storage
Upon receipt store plates at room temperature. Product shipped at ambient temperature.