Pierce™ NeutrAvidin™ Coated Plates
Thermo Scientific™
Pierce™ NeutrAvidin™ Coated Plates
Thermo Scientific™ Pierce NeutrAvidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of WellsFormatPlate Blocking
15116White96StandardSuperBlock™ Buffer
15217Black96StandardBlocker™ BSA Buffer
15117Black96StandardSuperBlock™ Buffer
15402Black384StandardSuperBlock™ Buffer
15128Clear96Strip PlateBlocker™ BSA Buffer
15127Clear96Strip PlateSuperBlock™ Buffer
15123Clear96StandardBlocker™ BSA Buffer
15129Clear96StandardSuperBlock™ Buffer
15400Clear384StandardSuperBlock™ Buffer
15216White96StandardBlocker™ BSA Buffer
15401White384StandardSuperBlock™ Buffer
Catalog number 15116
Price (INR)
30,653.00
Online offer
Ends: 31-Mar-2026
51,088.00
Save 20,435.00 (40%)
Each
Color:
White
No. of Wells:
96
Format:
Standard
Plate Blocking:
SuperBlock™ Buffer
Request bulk or custom format
Price (INR)
30,653.00
Online offer
Ends: 31-Mar-2026
51,088.00
Save 20,435.00 (40%)
Each
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.
Thermo Scientific NeutrAvidin Protein Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of NeutrAvidin Protein Coated Plates include:
  • White plates for chemiluminescent detection methods
  • Pre-blocked with Blocker BSA Buffer or SuperBlock Blocking Buffer
  • Easy and gentle immobilization of biotin-containing conjugates
  • Low nonspecific binding
  • No denaturing of the protein component of a conjugate upon binding
  • Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
  • Less expensive than streptavidin coated plates
  • Lowest nonspecific binding properties of all biotin-binding proteins
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Affinity
Detection MethodChemiluminescence
No. per Pack5 Plates
Plate BlockingSuperBlock™ Buffer
Product TypeMicroplate
Quantity5 plates
ColorWhite
FormatStandard
MaterialPolystyrene
No. of Wells96
Product LineNeutrAvidin Pierce
Surface TreatmentNeutrAvidin Protein
Unit SizeEach
Contents & Storage
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.