Thermo Scientific Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.FeaturesRead more
Have Questions?
Catalog Number
Quantity
15144
5 plates
Catalog number 15144
Price (INR)
31,392.00
Online offer
Ends: 31-Mar-2026
52,319.00
Save 20,927.00 (40%)
Each
Quantity:
5 plates
Request bulk or custom format
Price (INR)
31,392.00
Online offer
Ends: 31-Mar-2026
52,319.00
Save 20,927.00 (40%)
Each
Thermo Scientific Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
Features of Anti-GST Coated Plates:
• Antibody-based—plates are coated with mouse monoclonal anti-GST antibody • Sensitive—detects GST fusion proteins down to 7.5ng/well • Flexibility—binds native or denatured forms of GST; an alternative to glutathione-coated plates • Ready to use—pre-blocked with SuperBlock Blocking Buffer • Compatibility—Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding • Superior performance—improved signal-to-noise ratio relative to competitor's plates • Custom formats available—ask about our Custom Plate-Coating Service for coating 96-well and 384-well clear, white or black plates
Thermo Scientific Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use. Unlike glutathione coated plates, these anti-GST plates effectively bind both native and denatured forms of GST. In most cases, pre-purification of cell lysates is not necessary before screening and analysis of recombinant GST-tagged protein expression by ELISA using the plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Affinity
Detection MethodColorimetric
Packaging5 Plate
Plate BlockingSuperBlock™ Buffer
Product TypeDetection Plate, Immunoassay, ELISA
Quantity5 plates
ColorClear
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentAntibody
Unit SizeEach
Contents & Storage
Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature.