Thymolphthalein, pure, indicator
Thymolphthalein, pure, indicator
Thymolphthalein, pure, indicator
Thymolphthalein, pure, indicator
Acros Organics

Thymolphthalein, pure, indicator

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 151460500
also known as 15146
HSN 29322090
Price (INR)
12,120.00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,120.00
Each
Add to cart
Chemical Identifiers
CAS125-20-2
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H30O4
InChI KeyLDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Sulfated ash=<0.5 %
Visual transition intervalfrom pH 8.8 (colorless) to pH 10.5 (blue)
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only