Product Image
Gibco™

Kanamycin Sulphate (100X)

Kanamycin Sulfate is a water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus. Kanamycin acts by binding to the 30SRead more
Have Questions?
Catalog NumberQuantity
1516007020 x 100 mL
Catalog number 15160070
Price (INR)
-
Quantity:
20 x 100 mL
Kanamycin Sulfate is a water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus. Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria. Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria, and is used for the prevention of bacterial contamination of cell cultures. This product is provided as a 10 mg/ml stock solution. The recommended working concentration for cell culture is 100 μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
Gibco™ Kanamycin Sulfate is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Kanamycin Sulfate product made in our Grand Island facility (15160-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 X
Recommended StorageStorage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Sterile FilteredYes
Validated ApplicationBacterial Selection
Physical FormLiquid
Quantity20 x 100 mL
TypeKanamycin
Unit SizeEach