Isophthalaldehyde, 99%
Isophthalaldehyde, 99%
Isophthalaldehyde, 99%
Acros Organics

Isophthalaldehyde, 99%

CAS: 626-19-7 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 151740050
HSN 29122990
Price (INR)
4,687.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,687.00
Each
Chemical Identifiers
CAS626-19-7
IUPAC Namebenzene-1,3-dicarbaldehyde
Molecular FormulaC8H6O2
InChI KeyIZALUMVGBVKPJD-UHFFFAOYSA-N
SMILESO=CC1=CC(C=O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point88°C to 90°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only