1500 B2 4ft BSC only 230V NEMA UV
1500 B2 4ft BSC only 230V NEMA UV
1500 B2 4ft BSC only 230V NEMA UV
NSF listed Class II, Type B2 BSC incorporating SmartFlow intelligent compensation for downflow, energy efficient DC motors and line-of-sight graphical display and user controls for easy access.
Have Questions?
Catalog NumberItem DescriptionVoltage
1518-MAR1500 Series Class II, Type B2 BSC with UV, 4 ft (1.2m) nominal width, 230 VAC, 50/60 Hz NEMA power cord and internal receptacle230 VAC
Catalog number 1518-MAR
Price (INR)
-
Request A Quote
Item Description:
1500 Series Class II, Type B2 BSC with UV, 4 ft (1.2m) nominal width, 230 VAC, 50/60 Hz NEMA power cord and internal receptacle
Voltage:
230 VAC
1500 Series B2 BSC, NSF listed Class II, Type B2 BSC available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8m). Regional voltages selectable. Incorporates SmartFlow intelligent compensation for downflow, energy efficient DC motor, stainless steel interior and line-of-sight graphical display and user controls for easy access.
  • NSF listed Class II type B2 BSC
  • SmartFlow intelligent compensation for downflow
  • Energy efficient DC motor
  • Stainless steel interior
  • Line-of-sight graphical display and user controls for easy access
Specifications
Certifications/ComplianceNSF Class II Type B2 BSC
Energy Consumption (Operating Set Point)0.28 kw
Model1500 Series
Noise Level65 dBA
Power Consumption0.28 kw (Operating Set Point)
ReceptaclesTwo Duplex
TypeBiological Safety Cabinet
Volume (Metric) Exhaust1130 m3/hr. (with DIM), 1448 m3/hr. (with Traverse)
Item Description1500 Series Class II, Type B2 BSC with UV, 4 ft (1.2m) nominal width, 230 VAC, 50/60 Hz NEMA power cord and internal receptacle
Width (English) Exterior54.3 in.
Width (English) Interior48.5 in.
Width (Metric) Exterior1379 mm
Width (Metric) Interior1232 mm
Height (English)25.7 in. to 29.2 in. (Interior), 72.6 in. (Exterior)
Height (Metric)1844 mm (Exterior), 653 mm to 742 mm (Interior)
Voltage230 VAC
Amperage6 A
Unit SizeEach