2,2'-Biphenol, 99%
2,2'-Biphenol, 99%
2,2'-Biphenol, 99%
Acros Organics

2,2'-Biphenol, 99%

CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
151995000
also known as 15199
500 gPlastic bottle
Catalog number 151995000
also known as 15199
HSN 29072900
Price (INR)
31,317.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
31,317.00
Each
Chemical Identifiers
CAS1806-29-7
IUPAC Name[1,1'-biphenyl]-2,2'-diol
Molecular FormulaC12H10O2
InChI KeyIMHDGJOMLMDPJN-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 %
Melting point108°C to 111°C
Iron (Fe)=<15 ppm
Appearance (Color)Beige to light grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only