Product Image
Thermo Scientific™
96 Well Plate, Collagen I Coated Surface, Pack of 5
Color:Clear; Lid:With Lid
Have Questions?
Catalog NumberQuantity
152038Case of 20
Catalog number 152038
Price (INR)
45,151.00
Case of 20
Quantity:
Case of 20
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
  • Aseptically Manufactured
Specifications
Array8 x 12
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorClear
Description96F well plate, Collagen I, clear, with lid
No. per Case20
No. per Pack5
Packaging5/Pack, 20/Case
Product LineNunc
QuantityCase of 20
Surface TreatmentCollagen Type I
Well ShapeFlat
MaterialPolystyrene
No. of Wells96
SterilityAseptically Manufactured
TypeMicroplate
Volume (Metric) Well400 μL
Unit SizeCase of 20
Frequently asked questions (FAQs)
How many plates are contained in 96 Well Plate, Collagen I Coated Surface, Pack of 5 (Cat. No. 152038)?
96 Well Plate, Collagen I Coated Surface, Pack of 5 (Cat. No. 152038) provides 5 Cases with each Case containing 20 individual Nunc MicroWell Collagen Type I-coated 96-well microplates.
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.