2-Carboxybenzaldehyde, 99%
2-Carboxybenzaldehyde, 99%
2-Carboxybenzaldehyde, 99%
Acros Organics

2-Carboxybenzaldehyde, 99%

CAS: 119-67-5 | C8H6O3 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 152080250
also known as 15208
HSN 29183090
Price (INR)
4,166.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,166.00
Each
Chemical Identifiers
CAS119-67-5
IUPAC Name2-formylbenzoic acid
Molecular FormulaC8H6O3
InChI KeyDYNFCHNNOHNJFG-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point96°C to 99°C
Infrared spectrumConforms
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only