Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Acros Organics

Aminoacetaldehyde dimethyl acetal, 99%

CAS: 22483-09-6 | C4H11NO2 | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 153345000
also known as 15334
HSN 29225090
Price (INR)
56,220.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
56,220.00
Each
Chemical Identifiers
CAS22483-09-6
IUPAC Name2,2-dimethoxyethan-1-amine
Molecular FormulaC4H11NO2
InChI KeyQKWWDTYDYOFRJL-UHFFFAOYSA-N
SMILESCOC(CN)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only