Ramsey™ Grad-Line G176 Grade Control Paver Hydraulic Leveling Systems
Thermo Scientific™
Ramsey™ Grad-Line G176 Grade Control Paver Hydraulic Leveling Systems
The Thermo Scientific™ Ramsey™ Grad-Line G176 Grade Control has been discontinued; however, we will continue to provide technical support and spare parts on a limited and as available basis.
Have Questions?
Catalog NumberDescriptionAccuracyOutput Current
15377Grade control0.04 in. at wand; surface accuracy dependent on machines and conditionsAvailable to drive hydraulic valve coils; On/off: 2.5 A minimum; Proportional: 0.5 A maximum
Catalog number 15377
Price (INR)
-
Description:
Grade control
Accuracy:
0.04 in. at wand; surface accuracy dependent on machines and conditions
Output Current:
Available to drive hydraulic valve coils; On/off: 2.5 A minimum; Proportional: 0.5 A maximum
The Thermo Scientific™ Ramsey™ Grad-Line G176 Grade Control has been discontinued; however, we will continue to provide technical support and spare parts on a limited and as available basis.
Specifications
DescriptionGrade control
Output CurrentAvailable to drive hydraulic valve coils; On/off: 2.5 A minimum; Proportional: 0.5 A maximum
Temperature Range-7°C to +80°C
Accuracy0.04 in. at wand; surface accuracy dependent on machines and conditions
Resolution0.02 in. at wand
Unit Size-