2-Heptanone, 98%
2-Heptanone, 98%
2-Heptanone, 98%
Acros Organics

2-Heptanone, 98%

CAS: 110-43-0 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
100 mL
250 mL
1 L
Packaging:
View all
Plastic drum
Catalog number 154000051
Price (INR)
50,562.00
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (INR)
50,562.00
Each
Chemical Identifiers
CAS110-43-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4075 to 1.4095 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only