Product Image
Invitrogen™

UltraPure™ Sucrose

UltraPure™ Sucrose is suitable for forming density gradients for a variety of separation applications. It is used to separate RNAs,Read more
Have Questions?
Catalog NumberQuantity
155030225 kg
Catalog number 15503022
Price (INR)
23,593.00
Each
Quantity:
5 kg
Price (INR)
23,593.00
Each
UltraPure™ Sucrose is suitable for forming density gradients for a variety of separation applications. It is used to separate RNAs, to prepare bacteriophage λ-DNA arms, and to separate proteins. UltraPure™ Sucrose gradients typically are formed by use of gradient formers or by layering two or more sucrose solutions in varying amounts in a centrifuge tube.

Performance and Quality Testing: No protease activity detected.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialSucrose
For Use With (Application)Density Gradients - Separation Applications
Product LineUltraPure
PurityMolecular Biology Grade
Quantity5 kg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
FormPowder
Unit SizeEach
Contents & Storage
Store at room temperature.