1,3-Dibromopropane, 98%, stabilized
1,3-Dibromopropane, 98%, stabilized
1,3-Dibromopropane, 98%, stabilized
Acros Organics

1,3-Dibromopropane, 98%, stabilized

CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number 155045000
also known as 15504
HSN 29033920
Price (INR)
25,139.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
25,139.00
Each
Chemical Identifiers
CAS109-64-8
IUPAC Name1,3-dibromopropane
Molecular FormulaC3H6Br2
InChI KeyVEFLKXRACNJHOV-UHFFFAOYSA-N
SMILESBrCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Stabilizer400 ppm butylene oxide
Refractive index1.5220 to 1.5240 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only