UltraPure™ Buffer-Saturated Phenol
UltraPure™ Buffer-Saturated Phenol
Invitrogen™

UltraPure™ Buffer-Saturated Phenol

UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids. The reagent, which consists of UltraPure™ Phenol that hasRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
15513047400mL
15513039100 mL
Catalog number 15513047
Price (INR)
30,766.00
온라인 행사
Ends: 31-Mar-2026
45,578.00
Save 14,812.00 (32%)
Each
Add to cart
Quantity:
400mL
Price (INR)
30,766.00
온라인 행사
Ends: 31-Mar-2026
45,578.00
Save 14,812.00 (32%)
Each
Add to cart

UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids. The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4. When mixtures are extracted with UltraPure™ Buffer- Saturated Phenol, proteins are denatured and collect in the organic phase or at the interphase, while most nucleic acids remain in the aqueous phase. UltraPure™ Buffer-Saturated Phenol contains no preservatives. It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles.

Performance and Quality Testing: The appearance of the solution is evaluated at room temperature. No RNase or DNase activity detected.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPhenols
Packaging TypeBottle
Product LineUltraPure
PurityMolecular Biology Grade
Quantity400mL
Shipping ConditionRoom Temperature
FormLiquid
pH7.4
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

Do you have any information on DNA and RNA purification using phenol chloroform and alcohol precipitation?

Phenol extraction of proteins:

Phenol extraction is frequently used to remove proteins from nucleic acid solutions. A common protocol is to add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution, vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

Studies at Thermo Fisher Scientific have shown that the concentration of NaCl in the aqueous solution should not exceed 0.5 M for good recovery of DNA. Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether. After extraction, DNA is usually precipitated with ammonium acetate and ethanol as described in another protocol on this server. Ref. Karger, B. D. (1989) FOCUS 11, 14.

A good source of general information on the properties of phenol can be found in Wallace, Donald M. “Large and Small-Scale Phenol Extractions”. Methods in Enz. Volume 152 guide to Molecular Cloning Techniques. 1987. Academic Press, Inc. Berger and Kimmel, eds. Chap.4, pg 33-41.

(a) At pH 5 to 6 DNA is selectively retained in the organic phase and interphase, leaving RNA in the aqueous phase. Therefore a pH greater than 7 is needed if DNA is to be extracted.

(b) At pH values below 7.6, poly A+ RNA is lost to the organic phase if chloroform is not present.

(c) Optimal RNA yields in phenol extraction are obtained if the salt concentration is less than 0.15 M NaCl. Salt concentration in the sample is not a factor for larger DNA molecules.

To store RNA after extraction use DEPC-treated water.

What is the recommended protocol for phenol-extraction removal of proteins from nucleic acid containing solutions?

Below is a commonly used protocol:

(1) Add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution. Note: for RNA solutions, acid-phenol is recommended.

(2) Vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

(3) Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether.

(4) After extraction, DNA/RNA is usually precipitated with ammonium acetate and ethanol.

What is the optimal pH of the phenol:chloroform mixture for isolation of DNA?

Partitioning of the nucleic acids in phenol is pH dependent. At pH 7.0 or higher, both DNA and RNA partition into the aqueous phase. At an acidic pH (below 7.0) DNA is denatured and will move into the organic phase, but the RNA remains in the aqueous phase. The mixture should be adjusted to at least pH 7.4 for work with DNA.

Recently I came across a DNA purification technique, which uses urea during phenol extraction. What is the purpose of using urea?

Using urea during phenol extraction denatures the protein associated with the DNA and the proteins that bind the genomic DNA to the cell wall.