Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip
Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip
Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip
Thermo Scientific™
Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip
Thermo Scientific Pierce Streptavidin Coated High Sensitivity Plates,Black, 8-Well Strip are biotin-binding microplates that are coated to maximize capture efficiencyRead more
Have Questions?
Catalog NumberQuantity
155255 plates
Catalog number 15525
Price (INR)
-
Quantity:
5 plates
Request bulk or custom format
Thermo Scientific Pierce Streptavidin Coated High Sensitivity Plates,Black, 8-Well Strip are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
Features of Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip
Sensitive—detect down to 5 ng/mL of biotinylated IgG
Specific—low background with high signal-to-noise ratios
Robust—broad dynamic range
Ready to use—plates are supplied pre-blocked to save time
Thermo Scientific Pierce Coated Plates are coated using proprietary surface-activation technology to achieve maximum efficient coating of active streptavidin biotin-binding protein. The high sensitivity plates enable detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins.
Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low non-specific interactions.
Related Products
Pierce Streptavidin Coated High Sensitivity Plates, Clear, 8-Well Strip
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyHigh Sensitivity, Affinity
DescriptionPierce Streptavidin Coated High Sensitivity Plates, Black, 96-Well Strip
Detection MethodFluorescence
Packaging5 Plate
Plate BlockingBlocker™ BSA Buffer
Product TypeMicroplate
Quantity5 plates
ColorBlack
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentStreptavidin
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product is shipped at ambient temperature.