Pierce™ NeutrAvidin™ Coated High Sensitivity Plates, Clear, 8-Well Strip
Thermo Scientific™
Pierce™ NeutrAvidin™ Coated High Sensitivity Plates, Clear, 8-Well Strip
Thermo Scientific Pierce High Sensitivity NeutrAvidin Protein Coated Plates are biotin-binding microplates that are coated to maximize capture efficiency ofRead more
Have Questions?
Catalog NumberQuantity
155305 plates
Catalog number 15530
Price (INR)
46,677.00
Each
Quantity:
5 plates
Request bulk or custom format
Price (INR)
46,677.00
Each
Thermo Scientific Pierce High Sensitivity NeutrAvidin Protein Coated Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
Features of High Sensitivity NeutrAvidin Protein Coated Plates:
Sensitive—detect down to 5 ng/mL of biotinylated IgG
Specific—low background with high signal-to-noise ratios
Robust—broad dynamic range
Ready to use—plates are supplied pre-blocked to save time
These Thermo Scientific Pierce Coated Plates are coated using proprietary surface-activation technology to achieve maximum efficient coating of active streptavidin biotin-binding protein. The high sensitivity plates enable detection of low concentrations ( ng/mL) of biotinylated antibodies and other proteins. These plates make it easy to develop a colorimetric ELISA for samples that contain low levels of target.
Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions. Thermo Scientific NeutrAvidin Protein is a deglycosylated form of avidin, which has a near-neutral pI (6.3), resulting in even lower nonspecific interactions. NeutrAvidin Protein has the additional advantage of lacking the bacterial RYD sequence found on streptavidin, which eliminates off-target binding to the RGD binding domain of adhesion receptors present in a variety of cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyHigh Sensitivity, Affinity
DescriptionPierce NeutrAvidin Coated High Sensitivity Plates, Clear, 96-Well Strip
Detection MethodChemiluminescence
Packaging5 Plate
Plate BlockingBlocker™ BSA Buffer
Product TypeMicroplate
Quantity5 plates
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNeutrAvidin Pierce
Surface TreatmentNeutrAvidin Protein
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.