UltraPure™ 1 M Tris-HCl Buffers are pre-mixed, pH-adjusted, sterile-filtered solutions. Prepared as 1 M concentrates, these buffers can be dilutedRead more
Have Questions?
Catalog Number
Quantity
15568025
1 L
Catalog number 15568025
Price (INR)
6,978.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
6,978.00
Each
UltraPure™ 1 M Tris-HCl Buffers are pre-mixed, pH-adjusted, sterile-filtered solutions. Prepared as 1 M concentrates, these buffers can be diluted to the desired concentration and used in molecular biology or general biochemistry applications.
Performance and Quality Testing: No DNase, RNase, or protease activity detected
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 M
FormLiquid
Product LineUltraPure
Product TypeTris-HCl Buffer
Quantity1 L
Shipping ConditionRoom Temperature
pHpH 8.0
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
How should I store UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?
We recommend storing UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025) in the refrigerator (2-8 degrees C).
What alternative do you offer for UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?
We recommend Tris (1 M), pH 8.0, RNase‐free (Cat. No. AM9855G).