Fluorene, 98+%
Fluorene, 98+%
Fluorene, 98+%
Acros Organics

Fluorene, 98+%

CAS: 86-73-7 | C13H10 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 156135000
HSN 29029090
Price (INR)
27,625.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
27,625.00
Each
Chemical Identifiers
CAS86-73-7
IUPAC Name9H-fluorene
Molecular FormulaC13H10
InChI KeyNIHNNTQXNPWCJQ-UHFFFAOYSA-N
SMILESC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to light brown
GC>=98.0 %
Melting point113°C to 117°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only